Boiling flasks, flat bottom, short neck, standard taper joint, PYREX®
Supplier: Corning
4100-125 4100-1L 4100-250 4100-50 4100-500
CORN4100-125EA 30.9 SGD
CORN4100-125 CORN4100-1L CORN4100-250 CORN4100-50 CORN4100-500
Boiling flasks, flat bottom, short neck, standard taper joint, PYREX®
Flasks Flat Bottom Flasks
PYREX® borosilicate glass, clear, with short neck.
  • High thermal shock resistance
  • Chemically inert
  • Provide contamination free results
  • Long life
These flasks have full length outer standard tapered 24/40 joint. The normal working temperature is 230 °C.