Filtering flasks, graduated, heavy wall, PYREX®
Supplier: Corning
5341-1L 5341-250 5341-2L 5341-4L 5341-500
CORN5341-1LEA 920 SGD
CORN5341-1L CORN5341-250 CORN5341-2L CORN5341-4L CORN5341-500
Filtering flasks, graduated, heavy wall, PYREX®
Flasks Filter Flasks
PYREX® borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
These filtering flask have bent, poly tubulation in a grommet which acts as a shock absorber. Graduated to show approximate capacity.