Soxhlet extractor with standard taper joints, PYREX®
Supplier: Corning
3740-L 3740-M 3740-S 3740-XL
CORN3740-LEA 354 SGD
CORN3740-L CORN3740-M CORN3740-S CORN3740-XL
Soxhlet extractor with standard taper joints, PYREX®
Extractors Soxhlet and Soxhlet-Type Extractors
PYREX®, borosilicate glass.
For use with extraction apparatus with standard tapered ground joints at top and bottom. Bulb near the top of siphon tube aids siphoning action. Siphon tube protected from accidental damage by vapor tube. Capacity of extractor is measured to the top of the siphon tube.