Volumetric flasks, class A, with screw cap, PYREX®
Supplier: Corning
5650-100 5650-25 5650-50
CORN5650-100EA 453 SGD
CORN5650-100 CORN5650-25 CORN5650-50
Volumetric flasks, class A, with screw cap, PYREX®
Flasks Volumetric Flasks
Made of PYREX® borosilicate glass.
Flasks have an expanded open end to facilitate transfer of crystals, salts and powders. Standard GPI thread screw cap has an inert, cone-shaped, polyethylene liner for leakproof closure. Conforms to Class A capacity tolerances.