Distilling flasks, vertical type, 3 necks, [ST] joints, PYREX®
Supplier: Corning
4960-12L 4960-1L 4960-250 4960-2L 4960-500 4960-5L
CORN4960-12LEA 758 SGD
CORN4960-12L CORN4960-1L CORN4960-250 CORN4960-2L CORN4960-500 CORN4960-5L
Distilling flasks, vertical type, 3 necks, [ST] joints, PYREX®
Flasks Distillation Flasks
PYREX® borosilicate glass 3.3.
  • Excellent chemical resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
  • Made from heavy wall blanks to withstand severe service
Central neck and two parallel side necks. Matt white labelling area. Particularly suitable for use with a stirring device, condenser, or other apparatus as a unit.