Flasks, round bottom, glass, clear, standard tapered joint, one neck, PYREX® VISTA™
Supplier: Corning
70320-100 70320-1L 70320-250 70320-2L 70320-500 70320A-50
CORN70320-100EA 79.2 SGD
CORN70320-100 CORN70320-1L CORN70320-250 CORN70320-2L CORN70320-500 CORN70320A-50
Flasks, round bottom, glass, clear, standard tapered joint, one neck, PYREX® VISTA™
Flasks Round Bottom Flasks
PYREX® 7740 borosilicate glass, clear, with short neck.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
These flasks have full length outer standard tapered joint. Wall thickness is controlled to provide balance between thermal expansion and mechanical strength for shock resistance.
Certifications: ASTM E-1403