Distilling flask, vertical type, standard taper 24/40 joints, 3 necks, PYREX®
Supplier: Corning
4950-1L 4950-2L 4950-3L 4950-500 4950-5L
CORN4950-1LEA 170 SGD
CORN4950-1L CORN4950-2L CORN4950-3L CORN4950-500 CORN4950-5L
Distilling flask, vertical type, standard taper 24/40 joints, 3 necks, PYREX®
Flasks Distillation Flasks
PYREX® borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Central neck and two parallel side necks. Matt white labelling area. Three vertical neck flasks have two side necks with 24/40 outer standard taper joints and a center neck with a 24/40 standard taper joint. All of the necks are at the same height.