Serological pipettes, Ostwald-Folin, PYREX®

Supplier: Corning
7105-1 7105-2 7105-3 7105-5
CORN7105-1EA 267 SGD
CORN7105-1 CORN7105-2 CORN7105-3 CORN7105-5
Serological pipettes, Ostwald-Folin, PYREX®
Pipettes Serological Pipettes
This reusable blood chemistry pipettes were originally suggested by Folin for use in determining total nitrogen, urea and ammonia in urine, they are now generally used for blood serum tests.

  • Class A, non sterile
  • Calibrated to deliver (TD)
  • Colour-coded
  • Coloured graduations are enameled onto the glass
  • Round tip shape
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR