Volumetric flasks, class A, plastic coated, PYREXPLUS®
Supplier: Corning
65640-100 65640-1L 65640-200 65640-250 65640-2L 65640-500
CORN65640-100EA 952 SGD
CORN65640-100 CORN65640-1L CORN65640-200 CORN65640-250 CORN65640-2L CORN65640-500
Volumetric flasks, class A, plastic coated, PYREXPLUS®
Flasks Volumetric Flasks
PYREX® borosilicate glass, plastic coated, clear, with glass stoppers.
  • Resistant to thermal shock
  • Strength increased through machine-blown bodies
  • Sharp graduation line and permanent and large white block letters make the inscription easy to read
These flasks features a protective PVC coating for safety and long life, and it prevents glass from shattering and reduces spills.
Can be written on with any standard lab marker, permanent or water-soluble. Supplied with [ST] PYREX® stopper. It allows for a better hand grip and clamp hold.
Coating is autoclavable to 121 °C (250 °F). Do not heat above 121 °C (250 °F) moist heat or 110 °C (230 °F) dry heat.
Certifications: ASTM E-694, E-542, E-288
Caution: Caution: Do not place plastic-coated ware over direct heat or open flame.