Volumetric flasks, class A, PTFE stopper, PYREX®
Supplier: Corning
5644-10 5644-100 5644-1L 5644-200 5644-25 5644-250 5644-2L 5644-50 5644-500
CORN5644-10EA 588 SGD
CORN5644-10 CORN5644-100 CORN5644-1L CORN5644-200 CORN5644-25 CORN5644-250 CORN5644-2L CORN5644-50 CORN5644-500
Volumetric flasks, class A, PTFE stopper, PYREX®
Flasks Volumetric Flasks
These class A PYREX® volumetric flasks have machine-blown bodies to which are sealed heavy-beaded, heavy-tubing necks, tooled for PTFE color-coded keyhole stoppers.
  • Strength increased through machine-blown bodies
  • Sharp graduation line and permanent and large white block letters make the inscription easy to read
The graduation line is sharp and permanent. Large white block letters make the inscription easy to read. These flasks have been manufactured to Class A tolerances as established by ASTM E-694 for volumetric ware, ASTM E-542 for calibration ofMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable