Separating funnels, Squibb, glass [ST] stopper and [ST] glass key, Pyrex®
Supplier: Corning
6400-125 6400-1L 6400-250 6400-2L 6400-30 6400-4L 6400-500 6400-60 6400-6L
CORN6400-125EA 1120 SGD
CORN6400-125 CORN6400-1L CORN6400-250 CORN6400-2L CORN6400-30 CORN6400-4L CORN6400-500 CORN6400-60 CORN6400-6L
Separating funnels, Squibb, glass [ST] stopper and [ST] glass key, Pyrex®
Funnels Separating Funnels
PYREX®, borosilicate glass, with hollow glass stopper.
With socket and plain stem. With interchangeable glass key.
Stopcock bore, body opening and stem are carefully aligned for even liquid flow with fast cut-off. With smooth tooled tip.
Order Now
SPECIFICATIONS
Environmentally Preferable