Economy double metric scale cylinders, to contain, PYREX®
Supplier: Corning
3025-10 3025-100 3025-1L 3025-25 3025-250 3025-2L 3025-50 3025-500
CORN3025-10EA 464 SGD
CORN3025-10 CORN3025-100 CORN3025-1L CORN3025-25 CORN3025-250 CORN3025-2L CORN3025-50 CORN3025-500
Economy double metric scale cylinders, to contain, PYREX®
Cylinders Measuring Cylinders
Suitable for schools and labs requiring general accuracy only.
The double metric scale includes one reading down from top line and the other reading from bottom of cylinder to top line. Reduced tendency to tip due to relatively short height. Bumper guards not included.