Volumetric flasks, class A, glass, clear, glass [ST] stopper, PYREX®, VISTA™
Supplier: Corning
70640-10 70640-100 70640-1L 70640-200 70640-25 70640-250 70640-2L 70640-50 70640-500
CORN70640-10EA 189 SGD
CORN70640-10 CORN70640-100 CORN70640-1L CORN70640-200 CORN70640-25 CORN70640-250 CORN70640-2L CORN70640-50 CORN70640-500
Volumetric flasks, class A, glass, clear, glass [ST] stopper, PYREX®, VISTA™
Flasks Volumetric Flasks
PYREX® borosilicate glass, clear, with glass stoppers.
  • Flasks provide precise volume measurement
  • Flask neck tooled for No. 13 standard taper glass stoppers
  • Graduation lines are sharp and permanent
Vessels feature sealed necks tooled for standard taper stoppers and flat bottoms.
Certifications: ASTM E-694, E-544, E-288