Volumetric flasks, class A, glass, clear, glass [ST] stopper, PYREX® Lifetime Red™
Supplier: Corning
5660-100 5660-1L 5660-200 5660-25 5660-250 5660-2L 5660-50 5660-500
CORN5660-100EA 48.1 SGD
CORN5660-100 CORN5660-1L CORN5660-200 CORN5660-25 CORN5660-250 CORN5660-2L CORN5660-50 CORN5660-500
Volumetric flasks, class A, glass, clear, glass [ST] stopper, PYREX® Lifetime Red™
Flasks Volumetric Flasks
PYREX® borosilicate glass, clear, with glass stoppers.
These flasks have the capacity mark etched as a fine line through a narrow red band into clear glass.
Calibrated 'to contain' at 20 °C.
Certifications: ASTM E-694, E-542, E-288.
Order Now
SPECIFICATIONS
Environmentally Preferable