Bulb pipettes, PYREX® VISTA™, Class A
Supplier: Corning
70710-1 70710-10 70710-2 70710-5
CORN70710-1EA 60.9 SGD
CORN70710-1 CORN70710-10 CORN70710-2 CORN70710-5
Bulb pipettes, PYREX® VISTA™, Class A
Pipettes Bulb Pipettes
Reusable.
These bulb pipettes are manufactured to Class A capacity tolerances as indicated by ASTM E-969. The pipettes are colour-coded according to their capacity and feature coloured markings.