Measuring cylinders, tall form, borosilicate glass, PYREX®, Lifetime Red™
Supplier: Corning
3044-50 3044-100
CORN3044-50EA 740 SGD
CORN3044-50 CORN3044-100
Measuring cylinders, tall form, borosilicate glass, PYREX®, Lifetime Red™
Cylinders Measuring Cylinders
Borosilicate glass.
  • Calibrated to contain (TC)
  • Hexagonal base with graduations in white enamel
  • Tuttle type of low form for stability with double pourout