Serological pipettes, wide tip, PYREX®
Supplier: Corning
7087-10 7087-25 7087-5
CORN7087-10EA 276 SGD
CORN7087-10 CORN7087-25 CORN7087-5
Serological pipettes, wide tip, PYREX®
Pipettes Serological Pipettes
Glass, reusable. These serological pipettes are made with a large tip opening to allow more rapid intake and delivery of cell suspensions and viscous materials.
  • Calibrated to deliver (TD)
  • Coloured graduations are enameled on to the glass
  • Color-coded by size for easy identification and sorting
Order Now
SPECIFICATIONS
Environmentally Preferable