Erlenmeyer flasks, with standard ground joint, PYREX®
Supplier: Corning
5020-125 5020-1L 5020-25 5020-250 5020-50 5020-500
CORN5020-125EA 26.4 SGD
CORN5020-125 CORN5020-1L CORN5020-25 CORN5020-250 CORN5020-50 CORN5020-500
Erlenmeyer flasks, with standard ground joint, PYREX®
Flasks Erlenmeyer Flasks
PYREX® borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
These narrow mouth have standard taper stoppers for secure sealing. Their uniform wall thickness provides the proper balance between mechanical strength and thermal shock resistance. For convenience, these flasks are graduated to show approximate capacity. The 125 ml flasks are satisfactory for use as weighing bottles.
Order Now
SPECIFICATIONS
Environmentally Preferable