Erlenmeyer flasks, glass, clear, with standard ground joint, PYREX®
Supplier: Corning
5000-125 5000-250 5000-50 5000-500
CORN5000-125EA 37.5 SGD
CORN5000-125 CORN5000-250 CORN5000-50 CORN5000-500
Erlenmeyer flasks, glass, clear, with standard ground joint, PYREX®
Flasks Erlenmeyer Flasks
PYREX® borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
The uniform wall thickness of these flasks provide the proper balance between mechanical strength and thermal shock resistance. They are graduated to show approximate capacity and have an extra large marking space.
Certifications: ASTM E-123