Erlenmeyer flasks, narrow neck, PYREX® VISTA™
Supplier: Corning
70980-125 70980-1L 70980-25 70980-250 70980-50 70980-500
CORN70980-125EA 191 SGD
CORN70980-125 CORN70980-1L CORN70980-25 CORN70980-250 CORN70980-50 CORN70980-500
Erlenmeyer flasks, narrow neck, PYREX® VISTA™
Flasks Erlenmeyer Flasks
PYREX®, narrow neck.
  • Uniform wall thickness
  • Resistance to thermal shock
  • Approximate graduations
The uniform wall thickness to provide the proper balance between mechanical strength and thermal shock resistance.
these reusable flasks provides an extra-large blue marking space for easy identification. With Beaded top rims.
Certifications: ASTM E-1404 Type 1
Order Now
SPECIFICATIONS
Environmentally Preferable