Bell jars, with knob, PYREX®
Supplier: Corning
6885-140 6885-165 6885-222
CORN6885-140EA 896 SGD
CORN6885-140 CORN6885-165 CORN6885-222
Bell jars, with knob, PYREX®
Jars Bell Jars
These specially mold blown jars have a fine ground finish bottom flange to assure tight sealing with ground glass plates.
Caution: Do not use with heat, pressure or vacuum applications.