Adapters, glass, expansion, PYREX®
Supplier: Corning
8800-2419 8800-2924 8800-3424
CORN8800-2419EA 400 SGD
CORN8800-2419 CORN8800-2924 CORN8800-3424
Adapters, glass, expansion, PYREX®
Adapters Adapters, Conical Ground Joints
PYREX® glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Expansion adapter from small cone to large socket.