Paraffin Oil, J.T.Baker®
Supplier: Avantor
BAKRS894-00EA
222
SGD
BAKRS894-00
BAKRS894-07
Paraffin Oil, J.T.Baker®
Paraffin Oil
Boiling Pt:
300…500 °C (1013 hPa) Density: 0,86 g/cm³ (20 °C) Flash Pt: 215 °C (closed cup) |
CAS Number:
8012-95-1 EINECS: 232-384-2 Merck Index: 13,07093 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Identification (by IR) | Passes Test |
Product Information (not specifications): Saybolt Viscosity @ 100 | |
degrees F (sec).... 345 - 355 |