Reagent bottles, narrow neck, with standard taper stoppers, PYREX®
Supplier: Corning
1500-1L 1500-250 1500-2L 1500-500
CORN1500-1LEA 94.7 SGD
CORN1500-1L CORN1500-250 CORN1500-2L CORN1500-500
Reagent bottles, narrow neck, with standard taper stoppers, PYREX®
Bottles Glass Bottles
PYREX® borosilicate glass, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
These narrow mouth reagent storage bottles have sturdy ground necks with standard taper stoppers. Replacement stoppers are available.