Tissue grinders with glass pestles, PYREX®
Supplier: Corning
7725-13 7725-16 7725-19 7725-25
CORN7725-13EA 561 SGD
CORN7725-13 CORN7725-16 CORN7725-19 CORN7725-25
Tissue grinders with glass pestles, PYREX®
Homogenisers Tissue Grinders
Tissue grinders are used in preparing small samples for pathological and biochemical studies. All parts, including beaded rim and glass pestle, are ground to medium level for adequate abrasion and fine homogeneity.
Designed to be motor-driven
Caution: If used with motor-driven device, attachment must contain some sort of friction release clutch to prevent apparatus damage from piece of connective tissue or muscle. User should wear protective glove of sufficient thickness to prevent injury in event of glass components failure.
Order Now
SPECIFICATIONS
Environmentally Preferable