Melting point capillaries, PYREX®
Supplier: Corning
9530-1 9530-2 9530-3 9530-4
CORN9530-1EA 228 SGD
CORN9530-1 CORN9530-2 CORN9530-3 CORN9530-4
Melting point capillaries, PYREX®
Tubes Melting Point Tubes and Capillaries
Quality glassware at an affordable price.
  • Disposable
  • Available open-ended on both sides, or closed on one side
  • Made from no. 7740 glass
All tubes have a 0.2 to 0.25 mm wall range.