Separating funnels, Squibb, glass, PYREX®
Supplier: Corning
6406-125FO 6406-1L 6406-1LFO 6406-250 6406-250FO 6406-2L 6406-500 6406-500FO 6406-60 6406-60FO
CORN6406-125FOEA 263 SGD
CORN6406-125FO CORN6406-1L CORN6406-1LFO CORN6406-250 CORN6406-250FO CORN6406-2L CORN6406-500 CORN6406-500FO CORN6406-60 CORN6406-60FO
Separating funnels, Squibb, glass, PYREX®
Funnels Separating Funnels
PYREX® borosilicate glass, PTFE stopcock, HDPE stoppers.
These separatory funnels are sturdy and durable. The replaceable PTFE stopcock assembly features a simple screw thread locking nut and collar which ensure that the stopcock cannot fall out in use. The tip is replaceable. Its ground end provides a better grip. The complete funnel includes [ST] high density polyethylene stoppers of 7 mm Ø stem.
Ordering information: The replacement parts such as tip, stopcocks and funnel alone available and can be ordered separately.