Stoppers for conical ground joints, PYREX®
Supplier: Corning
7570N-19 7570N-24
CORN7570N-19EA 255 SGD
CORN7570N-19 CORN7570N-24
Stoppers for conical ground joints, PYREX®
Stoppers Glass Stoppers
Hollow, full length stopper closed at the bottom from PYREX®, borosilicate glass.
  • Interchangeable with standard taper joints
They are lightweight, but strong enough to keep chipping and breakage to a minimum. Barrel-shaped head is easy to clean and rotate, side grooves prevent slippage, and flat top allows stopper to stand on head.
Order Now
SPECIFICATIONS
Environmentally Preferable