Test tube racks
Supplier: NEW BRUNSWICK
M1289-0300 M1289-0100 M1289-0200 M1289-0400 M1289-0500 M1289-0600 M1289-0030 M1289-0003 M1289-0006 M1289-0001 M1289-0002 M1289-0005 M1289-0010 M1289-0020 M1289-0040 M1289-0050 M1289-0060
NEWBM1289-0300EA 0 SGD
NEWBM1289-0300 NEWBM1289-0100 NEWBM1289-0200 NEWBM1289-0400 NEWBM1289-0500 NEWBM1289-0600 NEWBM1289-0030 NEWBM1289-0003 NEWBM1289-0006 NEWBM1289-0001 NEWBM1289-0002 NEWBM1289-0005 NEWBM1289-0010 NEWBM1289-0020 NEWBM1289-0040 NEWBM1289-0050 NEWBM1289-0060
Test tube racks
Racks Tube Racks
Stainless steel
Test tube racks contains inserts to retain tubes and prevent them from spinning in the rack. Finger-tight knobs for adjusting the angle and releasing the rack from the base plate for easy transport to the bench.