Volumetric Flasks, Class A, Glass, Amber, TS Penny-head Glass Stopper
Supplier: HIRSCHMANN
264-G-100 264-G-1000
HIRS264-G-100EA 222 SGD
HIRS264-G-100 HIRS264-G-1000
Volumetric Flasks, Class A, Glass, Amber, TS Penny-head Glass Stopper
Flasks Volumetric Flasks
DURAN®, borosilicate glass, amber, with TS penny-head glass stopper.