Dropping bottles, glass, KIMBLE®
Supplier: KIMBLEKONTES
15040G-60 15040G-30
KIML15040G-60EA 155 SGD
KIML15040G-60 KIML15040G-30
Dropping bottles, glass, KIMBLE®
Bottles Dropping Bottles
Practical, comfortable to handle universal glass dropping bottles manufactured from amber soda-lime glass conforming to USP Type III and ASTM E438, Type II requirements.
  • Amber to protect from UV rays and light-sensitive substances
  • Good chemical and physical properties
  • For use in applications that do not involve high thermal stresses
Delivery information: Supplied with 20-400 GPI screw cap, bulb and glass dropper attached on the bottle.