Culture tubes, disposable, with screw thread finish, KIMBLE®
Supplier: KIMBLEKONTES
73760-16125 73770-13100 73770-16100 73770-16150
KIML73760-16125EA 1670 SGD
KIML73760-16125 KIML73770-13100 KIML73770-16100 KIML73770-16150
Culture tubes, disposable, with screw thread finish, KIMBLE®
Tubes Culture Tubes
Manufactured from 51 expansion borosilicate glass, KIMAX®.
  • Trays are shrink-wrapped in plastic film
  • No marking spots
  • Supplied without caps
These culture tubes suitable for tissue culture work and general bacteriological use.
Certifications: Glass conforming to USP Type I and ASTM E438, Type I, Class B requirements.
Order Now
SPECIFICATIONS
Environmentally Preferable