Erlenmeyer flasks, RAY-SORB®, KIMBLE®
Supplier: KIMBLEKONTES
26610-125 26610-500 26610-250
KIML26610-125EA 158 SGD
KIML26610-125 KIML26610-500 KIML26610-250
Erlenmeyer flasks, RAY-SORB®, KIMBLE®
Flasks Erlenmeyer Flasks
Manufactured from 33 expansion, low extractable borosilicate glass.
  • Without a capacity scale
  • With colour coded standard taper PTFE stopper
These flasks are processed to protect materials which are sensitive to the light of shorter wavelengths.
Designed from ASTM Specification E1404, Type II, Class II requirements. Glass conforming to to USP Type I and ASTM E438, Type I, Class A requirements, and then RAY-SORB® processed.
Order Now
SPECIFICATIONS
Environmentally Preferable