Weighing bottles, glass, KIMBLE®
Supplier: KIMBLEKONTES
15145-5060 15145-1550
KIML15145-5060EA 558 SGD
KIML15145-5060 KIML15145-1550
Weighing bottles, glass, KIMBLE®
Bottles Weighing Bottles
Manufactured from 33 expansion, low extractable borosilicate glass, KIMAX®.
  • Excellent chemical resistance
  • Excellent temperature resistance
These weighing bottles have a plug-style, closed bottom stopper that fits into an internally ground body. Ideal for determining the weight of solid samples.
Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
SPECIFICATIONS
Environmentally Preferable