Sample vials, with PTFE lined screw cap, KIMBLE®
Supplier: KIMBLEKONTES
60942A-40 60942A-8
KIML60942A-40EA 241 SGD
KIML60942A-40 KIML60942A-8
Sample vials, with PTFE lined screw cap, KIMBLE®
Vials
Sample vials made from clear KIMAX® KG-33 borosilicate glass.
  • Open top black PP closures attached
  • Closures with PTFE faced silicone septa
Certifications: Conforms USP Type I and ASTM E438, Type I, Class A requirements.