Weighing bottles, glass, low form, KIMBLE®
Supplier: KIMBLEKONTES
15165-7033 15165-5030
KIML15165-7033EA 754 SGD
KIML15165-7033 KIML15165-5030
Weighing bottles, glass, low form, KIMBLE®
Bottles Weighing Bottles
Manufactured from 33 expansion, low extractable borosilicate glass.
  • Excellent temperature resistance
  • Excellent chemical resistance
These weighing bottles have a plug-style glass stopper that creates a seal to secure contents and help to prevent spills and evaporation. Ideal for used in applications that require adding and removing substances to determine precise weight. The solid glass stopper with a short length standard taper joint fits into internally ground body.
Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
SPECIFICATIONS
Environmentally Preferable