Flasks acc. to kjeldahl, with standard ground joint, tooled neck, KIMBLE®
Supplier: KIMBLEKONTES
27400-500 27400-800
KIML27400-500EA 1300 SGD
KIML27400-500 KIML27400-800
Flasks acc. to kjeldahl, with standard ground joint, tooled neck, KIMBLE®
Flasks Kjeldahl Flasks
Manufactured from 33 expansion, low extractable borosilicate glass, KIMAX®.
  • Without a capacity scale
  • Flask necks tooled to accept rubber stoppers
Flasks have a reinforced bead at the top. These flasks are RAY-SORB® processed to protect materials which are sensitive to the light of shorter wavelengths.
Designed from ASTM specification E1404, Type II, Class II requirements. Glass conforming to to USP Type I and ASTM E438, Type I, Class A requirements, and then RAY-SORB® processed.