Büchner funnels, KIMBLE®
Supplier: KIMBLEKONTES
28400-303 28400-603
KIML28400-303EA 104 SGD
KIML28400-303 KIML28400-603
Büchner funnels, KIMBLE®
Funnels Filtration Funnels Büchner Funnels
Borosilicate glass, KIMAX®.
  • With KIMFLOW® fritted disc
  • With heavy walled stems
  • Available with marking spot
  • Beaded rims for extra strength
These funnels are used wherever the filter paper and membranes are unable to withstand the chemical attack of the filtrate.
Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.