Burettes, straight PTFE stopcock, class A, KIMBLE®
Supplier: KIMBLEKONTES
17027F-50 17027F-25 17027F-10
KIML17027F-50EA 509 SGD
KIML17027F-50 KIML17027F-25 KIML17027F-10
Burettes, straight PTFE stopcock, class A, KIMBLE®
Burettes
Borosilicate glass, class A.
  • Funnel fill style burette
  • Easy to read durable white ceramic enamel scale
  • Precision bore burette is permanently marked with an individual serial number and is traceable to NIST standards
  • Designed from ASTM specification, class A requirements
These burettes, marked with a serial number and certified, are used for general purpose titrations requiring traceable volumetric accuracy.
Delivery information: Supplied with a chemically resistant, self-lubricating PTFE stopcock plug, a KIM-KAP™ dust cap and a certificate of graduation accuracy.