Burettes, PTFE stopcock, educational grade, KIMBLE®
Supplier: KIMBLEKONTES
17121H-25 17121H-50
KIML17121H-25EA 187 SGD
KIML17121H-25 KIML17121H-50
Burettes, PTFE stopcock, educational grade, KIMBLE®
Burettes
Borosilicate glass.
  • Funnel fill style burette
  • Conforms to USP type I and ASTM specifications
These economical and versatile burettes are ideal for use in educational institutions and for many general laboratory procedures.
Note: The stopcock assemblies should be wetted before installation.
Delivery information: Supplied with PTFE threaded stopcock assembly, a precision ground tip and KIM-KAP™ dust cap.