Rods, glass, KIMBLE®
Supplier: KIMBLEKONTES
80700-1 80700-8
KIML80700-1EA 1150 SGD
KIML80700-1 KIML80700-8
Rods, glass, KIMBLE®
Solid Rods
Borosilicate glass.
  • Excellent chemical resistance
  • Excellent temperature resistance
Conforms to USP Type I, ASTM E438, Type I class A requirements.