Shell vials, with TITESEAL® plug style cap, KIMBLE®
Supplier: KIMBLEKONTES
60831D-830 60831D-843
KIML60831D-830EA 1250 SGD
KIML60831D-830 KIML60831D-843
Shell vials, with TITESEAL® plug style cap, KIMBLE®
Vials
These shell vials are made from clear KIMAX® N-51A borosilicate glass.
  • White PE plug style needle closure, unattached
  • Ideal for chromatography applications
Certifications: Glass conforms to USP Type I and ASTM E438, Type I, Class B requirements.
Order Now
SPECIFICATIONS
Environmentally Preferable