Bottles, wide neck, amber, with screw caps, with thread, KIMBLE®
Supplier: KIMBLEKONTES
5220438B 5220128B 5220128V-21 5220233V-21 5220438V-21 5220845V-21 5221653V-21 5223253V-21 5220438C-21 5220438C-22 5220438C-26 5220438V-25 5220438V-26
KIML5220438BEA 1150 SGD
KIML5220438B KIML5220128B KIML5220128V-21 KIML5220233V-21 KIML5220438V-21 KIML5220845V-21 KIML5221653V-21 KIML5223253V-21 KIML5220438C-21 KIML5220438C-22 KIML5220438C-26 KIML5220438V-25 KIML5220438V-26
Bottles, wide neck, amber, with screw caps, with thread, KIMBLE®
Bottles Glass Bottles
Soda-lime glass, amber.
  • Ideal for storage of solids
  • Wide-mouth design for efficient addition and removal of contents
Designed to protect contents from UV rays and ideal for light-sensitive products.
Conforms to USP Type III and ASTM E438, Type II requirements.