Flasks, round bottom, heavy wall, KIMBLE®
Supplier: KIMBLEKONTES
294000-0100 601000-0824
KIML294000-0100EA 63.9 SGD
KIML294000-0100 KIML601000-0824
Flasks, round bottom, heavy wall, KIMBLE®
Flasks Round Bottom Flasks
Manufactured from 33 expansion, low extractable borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
This flask is with a standard taper joint and made of heavy wall.
Certifications: Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.