Volumetric flasks, class A, glass, PTFE stopper, KIMBLE®
Supplier: KIMBLEKONTES
28014F-200 28014F-25 28014F-250 28014F-500
KIML28014F-200EA 974 SGD
KIML28014F-200 KIML28014F-25 KIML28014F-250 KIML28014F-500
Volumetric flasks, class A, glass, PTFE stopper, KIMBLE®
Flasks Volumetric Flasks
KIMAX®, borosilicate glass, with PTFE stopper.
  • With a marking spot
  • Standard taper stopper with a color-coded handle
These flasks are with a graduation ring blasted on the neck and are in a conventional flask shape.
Certifications: Calibrated to meet ASTM E-288, E-438, and glass conforms to USP type I, Class A Type I requirements.