Dishes, crystallising, KIMBLE®
Supplier: KIMBLEKONTES
23000-6035 23000-19100 23000-7050
KIML23000-6035EA 973 SGD
KIML23000-6035 KIML23000-19100 KIML23000-7050
Dishes, crystallising, KIMBLE®
Trays/Dishes/Pots Crystallising Dishes
Borosilicate glass, KIMAX®, without spout.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Ideal for storage and crystallization. Rims reinforced and fire-polished to reduce chipping.