Volumetric flasks, class A, micro, [ST] stopper, individually serialised, PYREX®
Supplier: Corning
5631-1 5631-10 5631-2 5631-25 5631-5
CORN5631-1EA 460 SGD
CORN5631-1 CORN5631-10 CORN5631-2 CORN5631-25 CORN5631-5
Volumetric flasks, class A, micro, [ST] stopper, individually serialised, PYREX®
Flasks Volumetric Flasks
PYREX® borosilicate glass, clear, with glass stoppers.
  • Hexagonal base to improve stability
  • Sharp and permanent markings
  • White markings for easy reading
  • With certificate of identification and capacity
  • Ideal for microchemical works
Certifications: ASTM E-237