Media/storage bottles, with screw cap, PYREXPLUS®
Supplier: Corning
61626-1L 61626-100 61626-250 61626-2L 61626-500
CORN61626-1LEA 424 SGD
CORN61626-1L CORN61626-100 CORN61626-250 CORN61626-2L CORN61626-500
Media/storage bottles, with screw cap, PYREXPLUS®
Bottles Cell Culture Bottles
PYREX® borosilicate glass, with screw cap.
  • Resistant to thermal shock
  • Teal enameled graduations and marking spot
  • Glass bead indicates full capacity line
These heavy duty bottles are ideal for storage, mixing and sampling. A protective PVC coating prevents glass from shattering and reduces spills. It includes green coloured PP plug seal cap with drip free pouring rings.
Autoclavable to 121 °C (249 °F) and have thread size: GL 45
Caution: Do not tighten caps immediately after autoclaving as the vacuum resulting from cooling can cause breakage. DoMore Product Information