Erlenmeyer flasks, wide neck,with screw cap, PYREX®
Supplier: Corning
4995-1L 4995-250 4995-2L 4995-4L 4995-500 4995-6L
CORN4995-1LEA 477 SGD
CORN4995-1L CORN4995-250 CORN4995-2L CORN4995-4L CORN4995-500 CORN4995-6L
Erlenmeyer flasks, wide neck,with screw cap, PYREX®
Flasks Erlenmeyer Flasks
Flasks made of PYREX® borosilicate glass, graduated.
  • Used as a vented culture flask when optional membrane cap is used
  • Convenient single size plug-seal cap
  • One-piece autoclavable orange polypropylene plug seal caps and drip-free pouring rings
For preparing and storing nutrient media and to use for larger volume serological pipets and ideal for use as a shaker flask.