Distilling flask, vertical necks, outer standard taper joints, 4-necks, PYREX®
Supplier: Corning
4935-1L 4935-2L 4935-5L
CORN4935-1LEA 345 SGD
CORN4935-1L CORN4935-2L CORN4935-5L
Distilling flask, vertical necks, outer standard taper joints, 4-necks, PYREX®
Flasks Distillation Flasks
PYREX® borosilicate glass, with vertical side necks, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Three vertical side necks placed 90° apart at the same height with 24/40 standard taper outer joints and a larger centre neck with a vertical 34/45 standard taper joint. Matt white labelling area.